3-(1,3-Oxazol-5-yl)benzoic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-(5-Oxazolyl)benzoic Acid

OR1404-1G OR1404-5G
77404-782EA 183.38 USD
77404-782 77404-790
3-(1,3-Oxazol-5-yl)benzoic acid 97%
3-(1,3-Oxazol-5-yl)benzoic acid
Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
Melting Pt: 258…260 °C
Storage Temperature: Ambient
MDL Number: MFCD06797481
CAS Number: 252928-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR