3-Benzylpyrrolidine

Supplier: APOLLO SCIENTIFIC
Warning

OR12550-1G OR12550-250MG
77404-364EA 591.94 USD
77404-364 77404-366
3-Benzylpyrrolidine
3-Benzylpyrrolidine
Formula: C₁₁H₁₅N
Storage Temperature: Refrigerator
MDL Number: MFCD01862537
CAS Number: 170304-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR