3-Phenylpyrrolidine

Supplier: APOLLO SCIENTIFIC
Warning

OR12549-100MG OR12549-1G OR12549-250MG
77404-352EA 21.26 USD
77404-352 77404-354 77404-356
3-Phenylpyrrolidine
3-Phenylpyrrolidine
Formula: C10H13N
Boiling Pt: 92…94 °C/2 mmHg
Density: 1.027 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD01838169
CAS Number: 936-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR