4-(Aminomethyl)-1-methyl-1H-imidazole 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR12312-1G OR12312-250MG OR12312-5G
77404-258EA 437.08 USD
77404-258 77404-260 77404-262
4-(Aminomethyl)-1-methyl-1H-imidazole 97%
4-(Aminomethyl)-1-methyl-1H-imidazole
Formula: C₅H₉N₃
Boiling Pt: 104…108 °C (0.2 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD06411536
CAS Number: 486414-83-9
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR