4-Methyl-2-(methylsulfanyl)-5-pyrimidinecarboxylic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Methyl-2-(methylthio)pyrimidine-5-carboxylic acid

OR12144-1G OR12144-500MG OR12144-5G
77404-208EA 78.46 USD
77404-208 77404-210 77404-212
4-Methyl-2-(methylsulfanyl)-5-pyrimidinecarboxylic acid 95%
4-Methyl-2-(methylsulfanyl)-5-pyrimidinecarboxylic acid
Formula: C₇H₈N₂O₂S
MW: 184.22 g/mol
Melting Pt: 169…171 °C
Storage Temperature: Ambient
MDL Number: MFCD02187391
CAS Number: 98276-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR