5-Aminolevulinic acid hydrochloride 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Amino-4-ketovaleric acid hydrochloride, 5-Amino-4-oxopentanoic acid hydrochloride, δ-Aminolevulinic Acid Hydrochloride

OR1490-1G OR1490-25G OR1490-5G
77405-164EA 21.32 USD
77405-164 77405-166 77405-168
5-Aminolevulinic acid hydrochloride 97%
5-Aminolevulinic acid hydrochloride
Formula: C₅H₁₀ClNO₃
MW: 167.59 g/mol
Melting Pt: 150 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00012869
CAS Number: 5451-09-2
EINECS: 226-679-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR