L-1-Naphthylalanine 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR14711-10G OR14711-5G
77405-084EA 13.39 USD
77405-084 77405-086 77405-088
L-1-Naphthylalanine 95%
L-1-Naphthylalanine
Formula: C₁₃H₁₃NO₂
Melting Pt: 254 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066086
CAS Number: 55516-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR