4-Methyl-D-phenylalanine 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: H-D-Phe(4-Me)-OH, H-p-Me-D-Phe-OH

OR14704-1G OR14704-5G
77405-078EA 21.26 USD
77405-078 77405-080
4-Methyl-D-phenylalanine 98%
4-Methyl-D-phenylalanine
Formula: C₁₀H₁₃NO₂
MW: 179.219 g/mol
Melting Pt: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00152137
CAS Number: 49759-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR