3-(Bromomethyl)benzaldehyde 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR1453-1G
OR1453-250MG
OR1453-5G
77405-008EA
93.74
USD
77405-008
77405-010
77405-012
3-(Bromomethyl)benzaldehyde 97%
3-(Bromomethyl)benzaldehyde
Formula:
C₈H₇BrO Melting Pt: 49…51 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797497 CAS Number: 82072-23-9 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...