(4S)-4-Benzyl-1,3-thiazolidine-2-thione 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (S)-4-Benzylthiazolidine-2-thione, (4S)-4-(Phenylmethyl)-2-thioxo-1,3-thiazolidine, (4S)-4-Benzyl-2-thioxo-1,3-thiazolidine

OR14521-10G OR14521-1G OR14521-5G
77404-998EA 390.2 USD
77404-998 77405-000 77405-002
(4S)-4-Benzyl-1,3-thiazolidine-2-thione 97%
(4S)-4-Benzyl-1,3-thiazolidine-2-thione
Formula: C₁₀H₁₁NS₂
MW: 209.331 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06658216
CAS Number: 171877-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR