8-Amino-2,4-dimethyl-2H-1,4-benzoxazin-3(4H)-one 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 8-Amino-3,4-dihydro-2,4-dimethyl-3-oxo-2H-1,4-benzoxazine

OR13821-1G OR13821-250MG
77404-742EA 529.78 USD
77404-742 77404-744
8-Amino-2,4-dimethyl-2H-1,4-benzoxazin-3(4H)-one 95%
8-Amino-2,4-dimethyl-2H-1,4-benzoxazin-3(4H)-one
Formula: C₁₀H₁₂N₂O₂
MW: 192.214 g/mol
Melting Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD09998375
CAS Number: 870064-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR