2-Amino-5-nitropyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR1164-100G OR1164-25G OR1164-500G
77403-978EA 28.52 USD
77403-978 77403-980 77403-984
2-Amino-5-nitropyridine
2-Amino-5-nitropyridine
Formula: C₅H₅N₃O₂
Melting Pt: 186…190 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006325
CAS Number: 4214-76-0
EINECS: 224-145-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR