5-Bromopyridine-2-carbonitrile 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR11382-25G OR11382-5G
77403-898EA 47.88 USD
77403-898 77403-900
5-Bromopyridine-2-carbonitrile 97%
5-Bromopyridine-2-carbonitrile
Formula: C₆H₃BrN₂
Boiling Pt: 100…110 °C/3 mmHg
Melting Pt: 124…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00234144
CAS Number: 97483-77-7
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR