4-Bromo-1,1,3,3,6-pentamethyl-7-nitro-2,3-dihydro-1H-inden-5-ylamine

Supplier: APOLLO SCIENTIFIC

Synonyms: 5-Amino-4-bromo-7-nitro-1,1,3,3,6-pentamethylindane

OR16156-1G OR16156-5G
77405-654EA 99.84 USD
77405-654 77405-656
4-Bromo-1,1,3,3,6-pentamethyl-7-nitro-2,3-dihydro-1H-inden-5-ylamine
4-Bromo-1,1,3,3,6-pentamethyl-7-nitro-2,3-dihydro-1H-inden-5-ylamine
Formula: C₁₄H₁₉BrN₂O₂
MW: 327.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100123
CAS Number: 1210378-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR