6-[(Tetrahydro-2H-pyran-4-yl)oxy]nicotinaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 6-[(Tetrahydro-2H-pyran-4-yl)oxy]pyridine-3-carboxaldehyde, 5-Formyl-2-[(tetrahydro-2H-pyran-4-yl)oxy]pyridine

OR14887-1G OR14887-250MG OR14887-50MG
77405-146EA 159.96 USD
77405-146 77405-148 77405-150
6-[(Tetrahydro-2H-pyran-4-yl)oxy]nicotinaldehyde 97%
6-[(Tetrahydro-2H-pyran-4-yl)oxy]nicotinaldehyde
Formula: C₁₁H₁₃NO₃
MW: 207.226 g/mol
Melting Pt: 66.5…68 °C
Storage Temperature: Ambient
MDL Number: MFCD09817493
CAS Number: 910036-95-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR