3-Imidazol-1-ylpropionic acid 96%

Supplier: APOLLO SCIENTIFIC
Warning

OR14498-1G OR14498-250MG
77404-976EA 206.82 USD
77404-976 77404-978
3-Imidazol-1-ylpropionic acid 96%
3-Imidazol-1-ylpropionic acid
Formula: C₆H₈N₂O₂
Melting Pt: 147…149 °C
Storage Temperature: Ambient
MDL Number: MFCD02731119
CAS Number: 18999-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR