3-(3-Hydroxypropyl)benzeneboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (3-(3-Hydroxypropyl)phenyl)boronic acid

OR13154-100MG OR13154-250MG
77404-556EA 59.1 USD
77404-556 77404-558 77404-560
3-(3-Hydroxypropyl)benzeneboronic acid
3-(3-Hydroxypropyl)benzeneboronic acid
Formula: C₉H₁₃BO₃
MW: 180.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04115650
CAS Number: 736989-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR