N-(2-Chloroethyl)-N'-(4-methoxyphenyl)urea

Supplier: APOLLO SCIENTIFIC

OR12061-1G OR12061-5G
77404-142EA 55.02 USD
77404-142 77404-144
N-(2-Chloroethyl)-N'-(4-methoxyphenyl)urea
N-(2-Chloroethyl)-N'-(4-methoxyphenyl)urea
Formula: C₁₀H₁₃ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD02671069
CAS Number: 65536-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR