6-Chloro-4-cyclopropyl-1H-quinazolin-2-one 95+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 6-Chloro-4-cyclopropylquinazolin-2(1H)-one

OR17533-100MG OR17533-250MG
77406-028EA 48.5 USD
77406-028 77406-030
6-Chloro-4-cyclopropyl-1H-quinazolin-2-one 95+%
6-Chloro-4-cyclopropyl-1H-quinazolin-2-one
Formula: C₁₁H₉ClN₂O
MW: 220.66 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11109434
CAS Number: 150878-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR