2-(Tributylstannyl)benzothiazole

Supplier: APOLLO SCIENTIFIC
Danger

OR17522-1G OR17522-250MG OR17522-500MG
77406-020EA 282.22 USD
77406-020 77406-022 77406-024
2-(Tributylstannyl)benzothiazole
2-(Tributylstannyl)benzothiazole
Formula: C₁₉H₃₁NSSn
Boiling Pt: 179 °C/1 mmHg
Density: 1.2023 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01319094
CAS Number: 105445-58-7
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR