(S)-3-(Aminomethyl)-1-boc-piperidine 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR15672-1G OR15672-25G OR15672-5G
77405-522EA 34.64 USD
77405-522 77405-524 77405-526
(S)-3-(Aminomethyl)-1-boc-piperidine 97%
(S)-3-(Aminomethyl)-1-Boc-piperidine
Formula: C₁₁H₂₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03839877
CAS Number: 140645-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR