4-(4-Ethylpiperazin-1-yl)phenylamine 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
4-(4-Ethylpiperazin-1-yl)aniline
OR15287-1G
OR15287-5G
77405-348EA
21.32
USD
77405-348
77405-350
4-(4-Ethylpiperazin-1-yl)phenylamine 98%
4-(4-Ethylpiperazin-1-yl)phenylamine
Formula:
C₁₂H₁₉N₃ MW: 205.3000 g/mol Melting Pt: 76…78 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00702246 CAS Number: 115619-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...