6-[(tert-Butoxycarbonyl)amino]nicotinic acid
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR15274-1G
OR15274-250MG
77405-342EA
123.28
USD
77405-342
77405-344
6-[(tert-Butoxycarbonyl)amino]nicotinic acid
6-[(tert-Butoxycarbonyl)amino]nicotinic acid
Formula:
C₁₁H₁₄N₂O₄ Melting Pt: 292…296 °C Storage Temperature: Ambient |
MDL Number:
MFCD02682388 CAS Number: 231958-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...