3-[4-(Hydroxymethyl)phenyl]pyridine 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (4-Pyridin-3-ylphenyl)methanol, (4-(Pyridin-3-yl)phenyl)methanol

OR1436-1G OR1436-250MG OR1436-5G
77404-932EA 172.06 USD
77404-932 77404-934 77404-936
3-[4-(Hydroxymethyl)phenyl]pyridine 97%
3-[4-(Hydroxymethyl)phenyl]pyridine
Formula: C₁₂H₁₁NO
MW: 185.222 g/mol
Melting Pt: 106…107 °C
Storage Temperature: Ambient
MDL Number: MFCD04116293
CAS Number: 217189-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR