3-(Chloromethyl)-5-methylisoxazole

Supplier: APOLLO SCIENTIFIC
Danger

OR1308-1G OR1308-250MG OR1308-5G
77404-508EA 66.22 USD
77404-508 77404-510 77404-512
3-(Chloromethyl)-5-methylisoxazole
3-(Chloromethyl)-5-methylisoxazole
Formula: C₅H₆ClNO
MW: 131.560 g/mol
Boiling Pt: 93 °C/15 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00221166
CAS Number: 35166-37-1
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR