3-Iodopyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR13049-100G OR13049-25G
77404-500EA 90.68 USD
77404-500 77404-502
3-Iodopyridine
3-Iodopyridine
Formula: C₅H₄IN
Melting Pt: 53…56 °C
Flash Pt: 106.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00023553
CAS Number: 1120-90-7
EINECS: 214-322-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR