N,N-Dimethyl-4-nitrobenzamide

Supplier: APOLLO SCIENTIFIC
Warning

OR111112-1G OR111112-5G
77403-660EA 57.06 USD
77403-660 77403-662
N,N-Dimethyl-4-nitrobenzamide
N,N-Dimethyl-4-nitrobenzamide
Formula: C₉H₁₀N₂O₃
MW: 194.187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00229078
CAS Number: 7291-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR