Benzotriazole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,2,3-Benzotriazole, 1H-1,2,3-Benzotriazole, 1H-Benzotriazole, 1H-Benzo[d][1,2,3]triazole

OR13222-250G OR13222-500G
77404-596EA 28.52 USD
77404-596 77404-598
Benzotriazole
Benzotriazole
Formula: C₆H₅N₃
Boiling Pt: 204 °C/15 mmHg
Melting Pt: 96…99 °C
Density: 1.36 g/cm³ (25 °C)
Flash Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00005699
CAS Number: 95-14-7
EINECS: 202-394-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR