5-Chloro-2-ethoxyphenylboronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR13120-250MG OR13120-25G OR13120-5G
77404-528EA 21.26 USD
77404-528 77404-530 77404-532
5-Chloro-2-ethoxyphenylboronic acid 98%
5-Chloro-2-ethoxyphenylboronic acid
Formula: C₈H₁₀BClO₃
Melting Pt: 120…124 °C
Storage Temperature: Ambient
MDL Number: MFCD03427055
CAS Number: 352534-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR