1-Cyclohexenylboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Cyclohex-1-en-1-ylboronic acid

OR13115-1G OR13115-25G OR13115-5G
77404-516EA 21.32 USD
77404-516 77404-518 77404-520
1-Cyclohexenylboronic acid
1-Cyclohexenylboronic acid
Formula: C₆H₁₁BO₂
MW: 125.96 g/mol
Melting Pt: 165 °C
Storage Temperature: Freezer
MDL Number: MFCD02179497
CAS Number: 89490-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR