2-Bromo-4-methylimidazole
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR1205-10G
OR1205-1G
OR1205-5G
77404-132EA
363.72
USD
77404-132
77404-134
77404-140
2-Bromo-4-methylimidazole
2-Bromo-4-methylimidazole
Formula:
C₄H₅BrN₂ Melting Pt: 110…114 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06659904 CAS Number: 23328-88-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...