4-Methoxyphenyl methylsulfonate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Methoxyphenyl mesylate, 4-[(Methylsulphonyl)oxy]anisole

OR11951-25G OR11951-5G
77404-084EA 737 USD
77404-084 77404-086
4-Methoxyphenyl methylsulfonate 98%
4-Methoxyphenyl methylsulfonate
Formula: C₈H₁₀O₄S
Storage Temperature: Ambient
MDL Number: MFCD01231360
CAS Number: 19013-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR