5-(Ethoxycarbonyl)-2-methoxybenzeneboronic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethyl 3-borono-4-methoxybenzoate, 5-(Ethoxycarbonyl)-2-methoxyphenylboronic acid, 2-Borono-4-(ethoxycarbonyl)anisole

OR11693-1G OR11693-250MG OR11693-5G
77404-010EA 167.08 USD
77404-010 77404-012 77404-014
5-(Ethoxycarbonyl)-2-methoxybenzeneboronic acid 95%
5-(Ethoxycarbonyl)-2-methoxybenzeneboronic acid
Formula: C₁₀H₁₃BO₅
Storage Temperature: Ambient
MDL Number: MFCD09800892
CAS Number: 957062-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR