Ethyl-2-bromo-4-oxazolecarboxylate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethyl 2-bromooxazole-4-carboxylate

OR11662-10G OR11662-1G OR11662-5G
77403-994EA 268.96 USD
77403-994 77403-996 77403-998
Ethyl-2-bromo-4-oxazolecarboxylate 98%
Ethyl 2-bromo-4-oxazolecarboxylate
Formula: C₆H₆BrNO₃
Melting Pt: 49…51 °C
Storage Temperature: Freezer
MDL Number: MFCD09801023
CAS Number: 460081-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR