6-Chloroimidazo[1,2-a]pyridine hydrochloride 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR11464-1G OR11464-5G
77403-922EA 22.28 USD
77403-922 77403-924
6-Chloroimidazo[1,2-a]pyridine hydrochloride 98%
6-Chloroimidazo[1,2-a]pyridine hydrochloride
Formula: C₇H₆Cl₂N₂
MW: 189.0400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09475876
CAS Number: 957035-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR