1,3-Dimethyl-1H-thieno[2,3-c]pyrazole-5-carbaldehyde
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR111192-1G
OR111192-5G
77403-682EA
374.83
USD
77403-682
77403-684
1,3-Dimethyl-1H-thieno[2,3-c]pyrazole-5-carbaldehyde
1,3-Dimethyl-1H-thieno[2,3-c]pyrazole-5-carbaldehyde
Formula:
C₈H₈N₂OS MW: 180.227 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03086154 CAS Number: 50880-19-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...