3-(Allyloxy)benzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR111184-10G OR111184-1G OR111184-5G
77403-674EA 192.4 USD
77403-674 77403-676 77403-678
3-(Allyloxy)benzaldehyde
3-(Allyloxy)benzaldehyde
Formula: C₁₀H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD04227161
CAS Number: 40359-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR