6-Methyl-1H-indazol-5-amine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Amino-6-methyl-1H-indazole

OR12141-1G OR12141-250MG
77404-200EA 104.16 USD
77404-200 77404-202
6-Methyl-1H-indazol-5-amine
6-Methyl-1H-indazol-5-amine
Formula: C₈H₉N₃
MW: 147.18 g/mol
Melting Pt: 215…217 °C
Storage Temperature: Freezer
MDL Number: MFCD03305372
CAS Number: 81115-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR