Methyl-2-acetyl-3-(dimethylamino)acrylate 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR12112-10G OR12112-1G OR12112-5G
77404-184EA 522.66 USD
77404-184 77404-186 77404-188
Methyl-2-acetyl-3-(dimethylamino)acrylate 95%
Methyl-2-acetyl-3-(dimethylamino)acrylate
Formula: C₈H₁₃NO₃
Melting Pt: 66…67 °C
Storage Temperature: Ambient
MDL Number: MFCD08689663
CAS Number: 203186-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR