4-Bromo-N-methylbenzamide 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR11557-1G OR11557-25G OR11557-5G
77403-948EA 21.36 USD
77403-948 77403-950 77403-952
4-Bromo-N-methylbenzamide 98%
4-Bromo-N-methylbenzamide
Formula: C₈H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD00462943
CAS Number: 27466-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR