5-Bromo-2-methoxynicotinonitrile 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR11516-1G OR11516-5G
77403-934EA 75.4 USD
77403-934 77403-936
5-Bromo-2-methoxynicotinonitrile 98%
5-Bromo-2-methoxynicotinonitrile
Formula: C₇H₅BrN₂O
Melting Pt: 110…115 °C
Storage Temperature: Ambient
MDL Number: MFCD09801047
CAS Number: 941294-54-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR