N-(9H-Purin-6-yl)benzamide 94%

Supplier: APOLLO SCIENTIFIC
Warning

OR11259-25G OR11259-5G
77403-862EA 29.54 USD
77403-862 77403-864
N-(9H-Purin-6-yl)benzamide 94%
N-(9H-Purin-6-yl)benzamide
Formula: C₁₂H₉N₅O
Melting Pt: 241…244 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00443632
CAS Number: 4005-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR