5-Cyano-2-thiopheneboronic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (5-Cyanothiophen-2-yl)boronic acid, 5-Cyanothiophene-2-boronic acid

OR11252-10G OR11252-1G OR11252-5G
77403-854EA 170.14 USD
77403-854 77403-856 77403-858
5-Cyano-2-thiopheneboronic acid 97%
5-Cyano-2-thiopheneboronic acid
Formula: C₅H₄BNO₂S
MW: 152.97 g/mol
Melting Pt: 125…128 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02094029
CAS Number: 305832-67-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR