Ethyl imidazo[1,2-a]pyridine-6-carboxylate 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR110709-1G OR110709-5G
77403-450EA 73.36 USD
77403-450 77403-452
Ethyl imidazo[1,2-a]pyridine-6-carboxylate 95%
Ethyl imidazo[1,2-a]pyridine-6-carboxylate
Formula: C₁₀H₁₀N₂O₂
MW: 190.20 g/mol
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD13176746
CAS Number: 158001-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR