Methyl 2-chloro-6-morpholin-4-ylisonicotinate

Supplier: APOLLO SCIENTIFIC

OR110252-1G OR110252-250MG
77403-178EA 194.6 USD
77403-178 77403-180
Methyl 2-chloro-6-morpholin-4-ylisonicotinate
Methyl 2-chloro-6-morpholin-4-ylisonicotinate
Formula: C₁₁H₁₃ClN₂O₃
MW: 256.686 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12545988
CAS Number: 1201675-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR