Ethyl-3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate

Supplier: APOLLO SCIENTIFIC

OR110198-1G OR110198-5G
77403-126EA 96.78 USD
77403-126 77403-128
Ethyl-3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₀H₈BrClN₂O₂
MW: 303.54
Melting Pt: 139…141 °C
Storage Temperature: Ambient
MDL Number: MFCD06496148
CAS Number: 861208-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR