Ethyl-3-amino-1-benzothiophene-2-carboxylate

Supplier: APOLLO SCIENTIFIC

Synonyms: Ethyl 3-aminobenzo[b]thiophene-2-carboxylate

OR110192-25G OR110192-5G
77403-118EA 273.04 USD
77403-118 77403-120
Ethyl-3-amino-1-benzothiophene-2-carboxylate
Ethyl-3-amino-1-benzothiophene-2-carboxylate
Formula: C₁₁H₁₁NO₂S
Melting Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00661903
CAS Number: 34761-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR