4,5-Dimethyl-1,3-thiazol-2-amine 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4,5-Dimethylthiazol-2-amine, 2-Amino-4,5-dimethyl-1,3-thiazole

OR14117-5G OR14117-1G OR14117-250MG
77404-814EA 28.44 USD
77404-814 77404-810 77404-812
4,5-Dimethyl-1,3-thiazol-2-amine 95%
4,5-Dimethyl-1,3-thiazol-2-amine
Formula: C₅H₈N₂S
MW: 128.2 g/mol
Melting Pt: 66…69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00462425
CAS Number: 2289-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR