Ethyl-2-methylthiazole-4-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethyl 2-methylthiazole-4-carboxylate

OR1301-10G OR1301-1G OR1301-5G
77404-486EA 33.62 USD
77404-486 77404-488 77404-490
Ethyl-2-methylthiazole-4-carboxylate
Ethyl 2-methylthiazole-4-carboxylate
Formula: C₇H₉NO₂S
MW: 171.22 g/mol
Melting Pt: 54…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00156148
CAS Number: 6436-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR