5-(Bromoacetyl)-3-phenylisoxazole 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Bromo-1-(3-phenylisoxazol-5-yl)ethan-1-one

OR1268-1G OR1268-250MG OR1268-5G
77404-404EA 69.28 USD
77404-404 77404-406 77404-408
5-(Bromoacetyl)-3-phenylisoxazole 95%
5-(Bromoacetyl)-3-phenylisoxazole
Formula: C₁₁H₈BrNO₂
MW: 266.091 g/mol
Melting Pt: 109…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00173898
CAS Number: 14731-14-7
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR